siRNA Details
Gene Symbol: Depdc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029916.1 | NP_001025087.1 | 2 | 216 | ||
XM_006234642.2 | XP_006234704.1 | 2 | 239 | |||
GenBank mRNA | BC097357.1 | - | 2 | 216 | 7442188 |
