siRNA Details
Gene Symbol: Dcdc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114621.1 | NP_001108093.1 | 10, 11 | 1255 | ||
XM_008762082.1 | XP_008760304.1 | 12, 13 | 2655 | |||
XM_008762083.1 | XP_008760305.1 | 6, 7 | 1387 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
