siRNA Details
Gene Symbol: Emc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106495.1 | NP_001099965.1 | 4 | 503 | ||
XM_006234704.2 | XP_006234766.1 | 5 | 1768 | |||
XM_008762088.1 | XP_008760310.1 | 4 | 1764 | |||
GenBank mRNA | FQ215850.1 | - | 4 | 443 | ||
FQ234005.1 | - | 4 | 443 |

More Information
Other Species Targeted

