siRNA Details
Gene Symbol: Arhgap11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168524.1 | NP_001161996.1 | 12 | 3080 | ||
GenBank mRNA | BC085123.1 | - | 2 | 1401 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Arhgap11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168524.1 | NP_001161996.1 | 12 | 3080 | ||
GenBank mRNA | BC085123.1 | - | 2 | 1401 |