siRNA Details
Gene Symbol: Vps18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106499.2 | NP_001099969.1 | 4 | 1614 | ||
GenBank mRNA | BC169083.1 | - | 4 | 1614 | 7936869 |

More Information
Other Species Targeted

Gene Ontology Categories:
