siRNA Details
Gene Symbol: Ndufaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106500.1 | NP_001099970.1 | 2 | 705 | ||
XM_006234737.1 | XP_006234799.1 | 2 | 585 | |||
XM_006234738.2 | XP_006234800.1 | 3 | 582 | |||
XM_006234739.2 | XP_006234801.1 | 2 | 573 | |||
XM_006234740.2 | XP_006234802.1 | 2 | 829 | |||
XM_006234741.2 | XP_006234803.1 | 2 | 729 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
