siRNA Details
Gene Symbol: Galk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013919.1 | NP_001013941.1 | 6 | 659 | ||
XM_006234904.2 | XP_006234966.1 | 5 | 1114 | |||
XM_006234906.2 | XP_006234968.1 | 5 | 619 | |||
GenBank mRNA | BC083716.1 | - | 6 | 659 | 7190694 |
