siRNA Details
Gene Symbol: ZBTB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143823.2 | NP_001137295.1 | 4 | 1704 | ||
NM_001243702.1 | NP_001230631.1 | 4 | 1372 | |||
NM_001243704.1 | NP_001230633.1 | 4 | 1363 | |||
NM_003409.4 | NP_003400.2 | 3 | 1674 | |||
GenBank mRNA | AK313236.1 | - | 3 | 1517 | ||
BC057282.1 | - | 4 | 1550 | |||
BC110519.2 | - | 2 | 1258 | 40033453 | ||
D89859.1 | - | 2 | 1279 | |||
Y12726.1 | - | 2 | 1246 |
