siRNA Details
Gene Symbol: Fam227b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013920.2 | NP_001013942.2 | 9 | 718 | ||
XM_006234909.2 | XP_006234971.1 | 9 | 641 | |||
XM_008762153.1 | XP_008760375.1 | 9 | 675 | |||
GenBank mRNA | BC079426.1 | - | 9 | 677 | 7114265 | |
BC089861.1 | - | 9 | 674 | 7114720 |
