siRNA Details
Gene Symbol: Cpxm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106511.1 | NP_001099981.1 | 14 | 2130 | ||
GenBank mRNA | FQ221560.1 | - | 2 | 152 |

More Information
Other Species Targeted

Gene Ontology Categories:
