siRNA Details
Gene Symbol: Pced1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012348.1 | NP_001012348.1 | 4 | 702 | ||
XM_006235015.2 | XP_006235077.1 | 6 | 1177 | |||
XM_006235016.2 | XP_006235078.1 | 7 | 2549 | |||
XM_006235017.2 | XP_006235079.1 | 6 | 1084 | |||
GenBank mRNA | BC089908.1 | - | 4 | 702 | 7304556 |

More Information
Other Species Targeted

