siRNA Details
Gene Symbol: Mcm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106514.1 | NP_001099984.1 | 7 | 943 | ||
XM_006235074.2 | XP_006235136.1 | 7 | 896 | |||
XM_008762163.1 | XP_008760385.1 | 7 | 928 |
