siRNA Details
Gene Symbol: Ankef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106516.1 | NP_001099986.1 | 9 | 2393 | ||
XM_008762252.1 | XP_008760474.1 | 9 | 4105 | |||
GenBank mRNA | BC160831.1 | - | 9 | 1543 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_175667, BC120825, AK029804.
