siRNA Details
Gene Symbol: Ndufaf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126371.1 | NP_001119843.1 | 8 | 788 | ||
GenBank mRNA | BC166551.1 | - | 8 | 788 | 7307233 | |
FQ213977.1 | - | 8 | 754 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_027093, AK009123, BC005630.
Gene Ontology Categories:
