siRNA Details
Gene Symbol: Dstnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751907.3 | XP_003751955.1 | 1 | 222 |
Gene Symbol: Dstn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033666.1 | NP_001028838.1 | 2 | 260 | ||
GenBank mRNA | CF111187.1 | - | 2 | 220 |

More Information
Other Species Targeted

Gene Ontology Categories:
