siRNA Details
Gene Symbol: Mgme1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009655.1 | NP_001009655.1 | 4, 5 | 994 | ||
XM_006235120.2 | XP_006235182.1 | 4, 5 | 1043 | |||
XM_006235121.1 | XP_006235183.1 | 3, 4 | 927 | |||
XM_006235122.2 | XP_006235184.1 | 4, 5 | 1060 | |||
XM_006235123.2 | XP_006235185.1 | 4, 5 | 1119 | |||
GenBank mRNA | BC087676.1 | - | 4, 5 | 994 | 7319345 |
