siRNA Details
Gene Symbol: Cst7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106523.1 | NP_001099993.1 | 2 | 268 | ||
XM_006235213.2 | XP_006235275.1 | 3 | 1541 | |||
XM_008762305.1 | XP_008760527.1 | 3 | 1541 | |||
XM_008762306.1 | XP_008760528.1 | 3 | 1538 |
