siRNA Details
Gene Symbol: Srxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762308.1 | XP_008760530.1 | 4 | 412 | ||
GenBank mRNA | AY325205.1 | - | 4 | 412 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
