siRNA Details
Gene Symbol: Tm9sf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025649.1 | NP_001020820.1 | 6 | 774 | ||
XM_006235284.2 | XP_006235346.1 | 6 | 792 | |||
GenBank mRNA | BC099133.1 | - | 6 | 774 | 7454474 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
