siRNA Details
Gene Symbol: Edem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004230.1 | NP_001004230.1 | 10 | 1195 | ||
GenBank mRNA | BC079029.1 | - | 10 | 1195 | 7114041 |

More Information
Other Species Targeted

Gene Ontology Categories:
