siRNA Details
Gene Symbol: Ergic3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106533.1 | NP_001100003.1 | 5 | 493 | ||
XM_006235314.1 | XP_006235376.1 | 5 | 612 | |||
GenBank mRNA | FQ213386.1 | - | 5 | 464 | ||
FQ230416.1 | - | 5 | 459 |
