siRNA Details
Gene Symbol: Ndrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013923.1 | NP_001013945.1 | 3 | 207 | ||
GenBank mRNA | BC078946.1 | - | 3 | 207 | 7109582 |
Gene Symbol: LOC679863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762423.1 | XP_008760645.1 | 3 | 252 |

More Information
Other Species Targeted

