siRNA Details
Gene Symbol: Bpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004079.1 | NP_001004079.1 | 11 | 1385 | ||
XM_008762323.1 | XP_008760545.1 | 11 | 1419 | |||
GenBank mRNA | BC079318.1 | - | 11 | 1385 | 7135061 |
