siRNA Details
Gene Symbol: Oser1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201560.2 | NP_963854.1 | 4 | 405 | ||
XM_006235528.2 | XP_006235590.1 | 4 | 1028 | |||
GenBank mRNA | AJ621831.1 | - | 4 | 409 | ||
BC082096.1 | - | 3 | 309 | 7134395 |
