siRNA Details
Gene Symbol: Ttpal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106537.1 | NP_001100007.1 | 4 | 776 | ||
XM_006235529.2 | XP_006235591.1 | 4 | 828 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
