siRNA Details
Gene Symbol: Ube2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106542.1 | NP_001100012.1 | 3 | 232 | ||
GenBank mRNA | FQ222472.1 | - | 3 | 223 | ||
FQ234128.1 | - | 3 | 226 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
