siRNA Details
Gene Symbol: Ctsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011959.1 | NP_001011959.1 | 10, 11 | 1246 | ||
XM_006235534.1 | XP_006235596.1 | 11, 12 | 1504 | |||
GenBank mRNA | BC078934.1 | - | 10, 11 | 1246 | 7108104 | |
FQ219992.1 | - | 3, 4 | 299 | |||
FQ229081.1 | - | 10, 11 | 1400 |
