siRNA Details
Gene Symbol: Zswim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011960.1 | NP_001011960.1 | 9 | 1672 | ||
GenBank mRNA | BC079431.1 | - | 9 | 1672 | 7114765 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
