siRNA Details
Gene Symbol: Ythdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024756.1 | NP_001019927.1 | 4 | 1293 | ||
GenBank mRNA | BC097360.1 | - | 4 | 1293 | 7442371 |

More Information
Other Species Targeted

Gene Ontology Categories:
