siRNA Details
Gene Symbol: Birc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762598.1 | XP_008760820.1 | 5 | 1495 | ||
XM_008762599.1 | XP_008760821.1 | 5 | 1495 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
