siRNA Details
Gene Symbol: Nkain4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235744.2 | XP_006235806.1 | 3, 4 | 376 | ||
XM_006235745.2 | XP_006235807.1 | 3, 4 | 2258 | |||
XM_008762496.1 | XP_008760718.1 | 4, 5 | 725 | |||
XM_008762497.1 | XP_008760719.1 | 4, 5 | 1553 | |||
XM_008762498.1 | XP_008760720.1 | 4, 5 | 528 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
