siRNA Details
Gene Symbol: Ppdpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009316.1 | NP_001009316.1 | 1 | 153 | ||
XM_006235746.2 | XP_006235808.1 | 2 | 305 | |||
XM_006235747.2 | XP_006235809.1 | 2 | 420 | |||
XM_006235748.2 | XP_006235810.1 | 2 | 205 | |||
GenBank mRNA | BC086948.1 | - | 1 | 153 | 7323892 | |
FQ211007.1 | - | 1 | 190 | |||
FQ227855.1 | - | 1 | 198 |

More Information
Other Species Targeted

Gene Ontology Categories:
