siRNA Details
Gene Symbol: CNBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK292119.1 | - | 3 | 310 | ||
AK314380.1 | - | 3 | 292 | |||
BC000288.2 | - | 3 | 309 | 2819226 | ||
BC093058.1 | - | 3 | 286 | 30721134 | ||
BT019613.1 | - | 2 | 161 | |||
DA606481.1 | - | 3 | 301 | |||
DB027227.1 | - | 3 | 347 | |||
M28372.1 | - | 3 | 263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
