siRNA Details
Gene Symbol: Dlx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100531.1 | NP_001094001.1 | 2 | 987 | ||
GenBank mRNA | BC167073.1 | - | 2 | 637 | 8364673 |

More Information
Other Species Targeted

Gene Ontology Categories:
