siRNA Details
Gene Symbol: Spopl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134447.1 | NP_001127919.1 | 9, 10 | 1152 | ||
GenBank mRNA | BC167106.1 | - | 9, 10 | 1152 | 7133920 |

More Information
Other Species Targeted

Gene Ontology Categories:
