siRNA Details
Gene Symbol: C8g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233581.2 | XP_006233643.1 | 1 | 509 | ||
GenBank mRNA | FQ210613.1 | - | 1 | 84 | ||
FQ212796.1 | - | 1 | 371 | |||
FQ218819.1 | - | 1 | 84 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
