siRNA Details
Gene Symbol: C8g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233581.2 | XP_006233643.1 | 1, 2 | 577 | ||
GenBank mRNA | FQ210613.1 | - | 1, 2 | 152 | ||
FQ212796.1 | - | 1, 2 | 439 | |||
FQ218819.1 | - | 1, 2 | 152 |

More Information
Other Species Targeted

Gene Ontology Categories:
