siRNA Details
Gene Symbol: Clic3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013080.2 | NP_001013098.2 | 3 | 244 | ||
XM_006233590.2 | XP_006233652.1 | 3 | 226 | |||
XM_008761584.1 | XP_008759806.1 | 2 | 932 | |||
XM_008761585.1 | XP_008759807.1 | 3 | 887 | |||
GenBank mRNA | BC089933.1 | - | 1 | 429 | ||
CF115110.1 | - | 3 | 244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
