siRNA Details
Gene Symbol: ZNF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006956.2 | NP_008887.2 | 6 | 1804 | ||
NM_016265.3 | NP_057349.2 | 5 | 1918 | |||
GenBank mRNA | AB021643.1 | - | 5, 6 | 1918 | ||
AF505656.1 | - | 5 | 1583 | |||
AK022691.1 | - | 1 | 768 | |||
AK293925.1 | - | 6 | 1564 | |||
AY500354.1 | - | 2 | 1220 | |||
BC051892.1 | - | 6 | 1792 | |||
BC137068.1 | - | 4 | 1410 | 9021065 | ||
BC137069.1 | - | 4 | 1410 | 9021066 | ||
BC144465.1 | - | 5 | 1463 | |||
BC154414.1 | - | 5 | 1906 | 40148925 | ||
BX640749.1 | - | 5 | 1329 | |||
X52334.1 | - | 1 | 140 |
