siRNA Details
Gene Symbol: Nacc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100533.1 | NP_001094003.1 | 2 | 493 | ||
XM_006233679.2 | XP_006233741.1 | 2 | 7592 | |||
XM_008761623.1 | XP_008759845.1 | 1 | 1696 | |||
GenBank mRNA | BC092608.1 | - | 1 | 297 | ||
CB720360.1 | - | 2 | 493 |
