siRNA Details
Gene Symbol: Nacc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100533.1 | NP_001094003.1 | 5, 6 | 1379 | ||
XM_006233679.2 | XP_006233741.1 | 5, 6 | 8478 | |||
XM_008761623.1 | XP_008759845.1 | 4, 5 | 2582 | |||
GenBank mRNA | BC092608.1 | - | 4, 5 | 1183 | ||
FQ210633.1 | - | 5, 6 | 788 |

More Information
Other Species Targeted

Gene Ontology Categories:
