siRNA Details
Gene Symbol: Slc2a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106562.1 | NP_001100032.1 | 9 | 1417 | ||
XM_006233757.2 | XP_006233819.1 | 9 | 1408 | |||
XM_008761634.1 | XP_008759856.1 | 9 | 1324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
