siRNA Details
Gene Symbol: Wdr34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005542.2 | NP_001005542.2 | 6 | 953 | ||
GenBank mRNA | BC082103.1 | - | 5 | 737 | 7134619 |

More Information
Other Species Targeted

