siRNA Details
Gene Symbol: Wdr34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005542.2 | NP_001005542.2 | 10 | 1635 | ||
GenBank mRNA | BC082103.1 | - | 9 | 1419 | 7134619 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wdr34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005542.2 | NP_001005542.2 | 10 | 1635 | ||
GenBank mRNA | BC082103.1 | - | 9 | 1419 | 7134619 |