siRNA Details
Gene Symbol: Asb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011963.1 | NP_001011963.1 | 2, 3 | 441 | ||
XM_006233912.2 | XP_006233974.1 | 2, 3 | 409 | |||
XM_008761641.1 | XP_008759863.1 | 1, 2 | 1175 | |||
XM_008761642.1 | XP_008759864.1 | 3, 4 | 420 | |||
GenBank mRNA | BC079099.1 | - | 2, 3 | 441 | 7124433 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
