siRNA Details
Gene Symbol: Asb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011963.1 | NP_001011963.1 | 4, 5 | 660 | ||
XM_006233912.2 | XP_006233974.1 | 4, 5 | 628 | |||
XM_008761641.1 | XP_008759863.1 | 3, 4 | 1394 | |||
GenBank mRNA | BC079099.1 | - | 4, 5 | 660 | 7124433 |
