siRNA Details
Gene Symbol: Prrc2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234005.1 | XP_006234067.1 | 10, 11 | 1320 | ||
XM_008761780.1 | XP_008760002.1 | 9, 10 | 1388 | |||
XM_008761781.1 | XP_008760003.1 | 10, 11 | 1451 | |||
XM_008761782.1 | XP_008760004.1 | 10, 11 | 1447 | |||
XM_008761783.1 | XP_008760005.1 | 9, 10 | 1388 | |||
XM_008761784.1 | XP_008760006.1 | 9, 10 | 1388 | |||
XM_008761785.1 | XP_008760007.1 | 9, 10 | 1385 | |||
XM_008761786.1 | XP_008760008.1 | 9, 10 | 1385 |
