siRNA Details
Gene Symbol: ZNF708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297560.1 | NP_001284489.1 | 4 | 920 | ||
NM_001297561.1 | NP_001284490.1 | 3 | 711 | |||
NM_021269.2 | NP_067092.2 | 4 | 836 | |||
XM_017027201.1 | XP_016882690.1 | 4 | 738 | |||
XM_017027202.1 | XP_016882691.1 | 4 | 861 | |||
XM_017027203.1 | XP_016882692.1 | 2 | 641 | |||
XM_017027204.1 | XP_016882693.1 | 4 | 653 | |||
GenBank mRNA | AK128184.1 | - | 4 | 795 | ||
AK131527.1 | - | 4 | 836 | |||
BX647526.1 | - | 4 | 906 |
