siRNA Details
Gene Symbol: Psmd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106569.2 | NP_001100039.2 | 8 | 1063 | ||
XM_006233969.2 | XP_006234031.1 | 8 | 1592 | |||
XM_006233970.2 | XP_006234032.1 | 9 | 1682 | |||
GenBank mRNA | BC166850.1 | - | 8 | 1045 | 9088649 |
