siRNA Details
Gene Symbol: Gsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004080.1 | NP_001004080.1 | 6, 7 | 908 | ||
XM_006234039.2 | XP_006234101.1 | 6, 7 | 806 | |||
XM_006234040.1 | XP_006234102.1 | 7, 8 | 926 | |||
GenBank mRNA | BC079472.1 | - | 6, 7 | 908 | 7122783 | |
FQ223877.1 | - | 7, 8 | 931 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_146120, NM_001206367, NM_001206368, NM_001206369, AK150359, and others.
Gene Ontology Categories:
